Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193191.2 | NP_001180120.2 | 1 - 2 | 421 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |