Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195013.1 | NP_001181942.1 | 9 - 10 | 1592 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |