Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANP32A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195019.1 | NP_001181948.1 | 2 - 3 | 347 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |