Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034552.1 | NP_001029724.1 | 1 - 2 | 26 | 61 | |
GenBank mRNA | BC146146.1 | - | 1 - 2 | 132 | 61 | |
BT021655.1 | - | 1 - 2 | 26 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |