Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14H8orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199131.1 | NP_001186060.1 | 12 - 13 | 1760 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |