Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204119.1 | NP_001191048.1 | 1 - 2 | 39 | 66 | |
NM_001371509.1 | NP_001358438.1 | 1 - 2 | 263 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |