Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204119.1 | NP_001191048.1 | 2 - 3 | 122 | 73 | |
NM_001371509.1 | NP_001358438.1 | 2 - 3 | 346 | 73 | ||
GenBank mRNA | DN283780.1 | - | 1 - 2 | 45 | 73 | |
DN511821.1 | - | 1 - 2 | 45 | 73 | ||
DN546048.1 | - | 1 - 2 | 45 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |