Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205320.1 | NP_001192249.1 | 2 - 3 | 233 | 110 | |
GenBank mRNA | EV612341.1 | - | 2 - 3 | 233 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |