Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFN-TAU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245936.1 | NP_001232865.1 | 1 | 423 | 93 | |
GenBank mRNA | HQ235019.1 | - | 1 | 423 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |