Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046432.1 | NP_001039897.1 | 1 | 1218 | 85 | |
GenBank mRNA | BC114052.1 | - | 1 | 1218 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |