Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270317.4 | NP_001257246.2 | 23 - 24 | 6719 | 69 | |
NM_001373583.1 | NP_001360775.1 | 23 - 24 | 6686 | 69 | ||
NM_001381151.1 | NP_001368758.1 | 23 - 24 | 6702 | 69 | ||
NM_001381152.1 | NP_001368759.1 | 23 - 24 | 6702 | 69 | ||
NM_001381153.1 | NP_001368756.1 | 23 - 24 | 6669 | 69 | ||
NM_001381154.1 | NP_001368757.1 | 23 - 24 | 6669 | 69 | ||
NM_001381155.1 | NP_001368146.1 | 23 - 24 | 6696 | 69 | ||
NM_001381156.2 | NP_001368760.1 | 23 - 24 | 6692 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |