Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270317.4 | NP_001257246.2 | 8 - 9 | 2258 | 73 | |
NM_001373583.1 | NP_001360775.1 | 8 - 9 | 2258 | 73 | ||
NM_001381155.1 | NP_001368146.1 | 8 - 9 | 2235 | 73 | ||
NM_001381156.2 | NP_001368760.1 | 8 - 9 | 2264 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |