Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syg-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322663.4 | NP_001309674.1 | 9 - 10 | 1308 | 73 | |
NM_001322664.3 | NP_001309467.1 | 9 - 10 | 1308 | 73 | ||
NM_001322665.4 | NP_001309678.1 | 4 - 5 | 380 | 73 | ||
NM_001383675.1 | 9 - 10 | 1301 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |