Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csq-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078037.7 | NP_510438.1 | 8 - 9 | 1053 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |