Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078052.5 | NP_510453.1 | 1 - 2 | 145 | 92 | |
GenBank mRNA | AF052060.1 | - | 1 - 2 | 175 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |