Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373610.3 | NP_001360030.1 | 2 - 3 | 345 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |