Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sod-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078363.8 | NP_510764.1 | 4 - 5 | 663 | 69 | |
GenBank mRNA | X85790.1 | - | 623 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |