Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acdh-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078388.5 | NP_510789.1 | 1 - 2 | 426 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |