Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377804.1 | NP_001364770.1 | 13 - 14 | 2325 | 65 | |
NM_001381436.2 | NP_001367838.1 | 12 - 13 | 2290 | 65 | ||
NR_131363.1 | 12 - 13 | 2260 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |