Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sop-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306444.3 | NP_001293373.1 | 4 - 5 | 523 | 65 | |
NM_001377813.3 | NP_001364767.1 | 4 - 5 | 522 | 65 | ||
NM_001377816.2 | NP_001364504.1 | 4 - 5 | 526 | 65 | ||
NM_170837.5 | NP_740821.1 | 4 - 5 | 526 | 65 | ||
GenBank mRNA | AF308860.1 | - | 4 - 5 | 525 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |