Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sop-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306444.3 | NP_001293373.1 | 9 - 10 | 2307 | 97 | |
NM_001306445.3 | NP_001293374.1 | 4 - 5 | 1340 | 97 | ||
NM_001306446.3 | NP_001293375.1 | 4 - 5 | 1340 | 97 | ||
NM_001377813.3 | NP_001364767.1 | 9 - 10 | 2306 | 97 | ||
NM_001377816.2 | NP_001364504.1 | 9 - 10 | 2310 | 97 | ||
NM_170837.5 | NP_740821.1 | 9 - 10 | 2310 | 97 | ||
GenBank mRNA | AF308860.1 | - | 9 - 10 | 2309 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |