Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001374886.2 | NP_001361963.1 | 14 - 15 | 4226 | 145 | |
NM_001380120.2 | NP_001367609.1 | 14 - 15 | 4241 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |