Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aman-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001374889.1 | NP_001361919.1 | 3 - 4 | 405 | 181 | |
NM_001374890.2 | NP_001362173.1 | 4 | 412 | 181 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |