Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313565.3 | NP_001300494.1 | 4 | 468 | 103 | |
NM_001313566.3 | NP_001300495.1 | 4 | 468 | 103 | ||
NM_001313568.3 | NP_001300497.1 | 3 - 4 | 366 | 103 | ||
NM_001313569.3 | NP_001300498.1 | 3 - 4 | 366 | 103 | ||
NM_001313570.3 | NP_001300499.1 | 3 - 4 | 366 | 103 | ||
NM_001313571.3 | NP_001300500.1 | 3 - 4 | 366 | 103 | ||
NM_001313575.3 | NP_001300504.1 | 1 - 2 | 45 | 103 | ||
NM_001381461.1 | NP_001368508.1 | 4 - 5 | 468 | 103 | ||
NM_001381462.1 | NP_001368509.1 | 1 - 2 | 45 | 103 | ||
NM_001381463.1 | NP_001367420.1 | 4 - 5 | 541 | 103 | ||
NM_001383871.1 | 4 - 5 | 512 | 103 | |||
NM_001383873.1 | 4 - 5 | 534 | 103 | |||
NM_171011.4 | NP_741012.1 | 4 - 5 | 528 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |