Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373741.2 | NP_001360471.1 | 10 - 11 | 3346 | 84 | |
NM_001373742.2 | NP_001360472.1 | 10 - 11 | 3346 | 84 | ||
NM_001373743.2 | NP_001360024.1 | 10 - 11 | 3346 | 84 | ||
NM_001373744.2 | NP_001360023.1 | 10 - 11 | 3348 | 84 | ||
NR_151568.1 | 10 - 11 | 3346 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |