Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171035.6 | NP_741041.1 | 13 - 14 | 1778 | 83 | |
NM_171036.6 | NP_741042.1 | 8 - 9 | 1351 | 83 | ||
GenBank mRNA | LC026472.1 | - | 12 - 13 | 1722 | 83 | |
LC026473.1 | - | 8 - 9 | 1236 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |