Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pll-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171059.6 | NP_741067.1 | 8 - 9 | 1092 | 90 | |
NM_171060.5 | NP_741068.1 | 8 - 9 | 1120 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |