Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gar-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027422.4 | NP_001022593.1 | 2 - 3 | 349 | 77 | |
NM_001027423.6 | NP_001022594.1 | 2 - 3 | 361 | 77 | ||
NM_001027425.5 | NP_001022596.1 | 2 - 3 | 306 | 77 | ||
NM_001027426.4 | NP_001022597.1 | 2 - 3 | 363 | 77 | ||
NM_001379757.2 | NP_001368076.1 | 4 - 5 | 485 | 77 | ||
GenBank mRNA | AF272738.1 | - | 2 - 3 | 306 | 77 | |
AY053364.1 | - | 2 - 3 | 306 | 77 | ||
AY053365.1 | - | 2 - 3 | 306 | 77 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |