Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gar-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027422.4 | NP_001022593.1 | 4 - 5 | 659 | 75 | |
NM_001027423.6 | NP_001022594.1 | 4 - 5 | 671 | 75 | ||
NM_001027425.5 | NP_001022596.1 | 4 - 5 | 616 | 75 | ||
NM_001379757.2 | NP_001368076.1 | 6 - 7 | 795 | 75 | ||
GenBank mRNA | AF272738.1 | - | 4 - 5 | 616 | 75 | |
AY053364.1 | - | 4 - 5 | 616 | 75 | ||
AY053365.1 | - | 4 - 5 | 616 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |