Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lim-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066082.5 | NP_498483.1 | 6 - 7 | 1293 | 72 | |
NM_066083.7 | NP_498484.1 | 6 - 7 | 1526 | 72 | ||
NM_171173.4 | NP_741201.1 | 9 - 10 | 1521 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |