Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276777.3 | NP_001263706.1 | 2 - 3 | 334 | 96 | |
NM_001377564.1 | NP_001364690.1 | 1 - 2 | 77 | 96 | ||
NM_001377565.2 | NP_001364691.1 | 1 - 2 | 77 | 96 | ||
NM_066127.4 | NP_498528.1 | 1 - 2 | 77 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |