Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171180.5 | NP_741210.3 | 9 - 10 | 2033 | 104 | |
NM_171880.6 | NP_741211.3 | 9 - 10 | 2035 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |