Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027810.5 | NP_001022981.1 | 19 - 20 | 3430 | 89 | |
NM_001129275.4 | NP_001122747.1 | 9 - 10 | 1533 | 89 | ||
NM_001129276.3 | NP_001122748.1 | 20 - 21 | 3514 | 89 | ||
NM_001268079.4 | NP_001255008.1 | 20 - 21 | 3529 | 89 | ||
NM_001268080.3 | NP_001255009.1 | 15 - 16 | 2619 | 89 | ||
NM_001306679.3 | NP_001293608.1 | 13 - 14 | 2415 | 89 | ||
NM_001306687.3 | NP_001293616.1 | 14 - 15 | 2433 | 89 | ||
NM_001306688.3 | NP_001293617.1 | 10 - 11 | 1617 | 89 | ||
NM_001382965.1 | 22 - 23 | 3835 | 89 | |||
NM_171221.7 | NP_741263.3 | 21 - 22 | 3521 | 89 | ||
GenBank mRNA | AF424978.1 | - | 19 - 20 | 3366 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |