Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027641.4 | NP_001022812.1 | 11 - 12 | 2316 | 107 | |
GenBank mRNA | AF497513.1 | - | 11 - 12 | 2303 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |