Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028052.5 | NP_001023223.1 | 7 - 8 | 1166 | 119 | |
NM_001028053.5 | NP_001023224.1 | 2 - 3 | 258 | 119 | ||
NM_001380068.1 | NP_001368271.1 | 2 - 3 | 172 | 119 | ||
NM_067740.6 | NP_500141.1 | 7 - 8 | 1060 | 119 | ||
NM_067741.5 | NP_500142.1 | 7 - 8 | 931 | 119 | ||
NM_171279.6 | NP_741329.1 | 7 - 8 | 967 | 119 |
Gene Symbol: ccm-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380067.2 | NP_001367894.1 | 16 - 17 | 1958 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |