Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038349.4 | NP_001033438.1 | 9 - 10 | 2376 | 85 | |
NM_001380147.1 | NP_001368216.1 | 5 - 6 | 1851 | 85 | ||
NM_001380148.1 | NP_001368210.1 | 3 - 4 | 1383 | 85 | ||
GenBank mRNA | Z14146.1 | - | 2441 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |