Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027998.6 | NP_001023169.1 | 6 - 7 | 1136 | 147 | |
NM_171382.4 | NP_741445.1 | 6 - 7 | 1152 | 147 | ||
GenBank mRNA | AF083226.1 | - | 6 - 7 | 1140 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |