Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059408.5 | NP_491809.2 | 10 - 11 | 2026 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |