Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bli-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026372.7 | NP_001021543.1 | 12 - 13 | 2136 | 70 | |
NM_001380517.2 | NP_001367786.1 | 12 - 13 | 2118 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |