Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snt-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059452.8 | NP_491853.2 | 7 - 8 | 1124 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |