Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059454.4 | NP_491855.1 | 12 - 13 | 4906 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |