Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025858.4 | NP_001021029.1 | 12 - 13 | 2503 | 109 | |
NM_001306351.3 | NP_001293280.1 | 10 - 11 | 1841 | 109 | ||
NM_001306352.3 | NP_001293281.1 | 8 - 9 | 1394 | 109 | ||
NM_059539.6 | NP_491940.2 | 11 - 12 | 2316 | 109 | ||
GenBank mRNA | AF170425.1 | - | 12 - 13 | 2272 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |