Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cye-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025856.4 | NP_001021027.1 | 6 - 7 | 1144 | 90 | |
NM_001025857.4 | NP_001021028.1 | 6 - 7 | 1142 | 90 | ||
GenBank mRNA | AF058331.1 | - | 6 - 7 | 1164 | 90 | |
AF520616.1 | - | 6 - 7 | 1166 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |