Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cye-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025856.4 | NP_001021027.1 | 7 - 8 | 1528 | 153 | |
NM_001025857.4 | NP_001021028.1 | 7 - 8 | 1526 | 153 | ||
GenBank mRNA | AF058331.1 | - | 7 - 8 | 1548 | 153 | |
AF520616.1 | - | 7 - 8 | 1550 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |