Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpf-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059555.2 | NP_491956.1 | 13 - 14 | 1942 | 89 | |
NM_059556.5 | NP_491957.1 | 13 - 14 | 1930 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |