Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpf-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059555.2 | NP_491956.1 | 19 - 20 | 2657 | 150 | |
NM_059556.5 | NP_491957.1 | 19 - 20 | 2645 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |