Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ekl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059608.5 | NP_492009.1 | 7 - 8 | 1488 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |