Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ztf-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129030.3 | NP_001122502.1 | 3 - 4 | 370 | 106 | |
NM_059651.6 | NP_492052.2 | 3 - 4 | 239 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |