Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rde-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026227.3 | NP_001021398.1 | 5 - 6 | 693 | 89 | |
NM_001026228.4 | NP_001021399.1 | 5 - 6 | 722 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |