Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sol-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059687.7 | NP_492088.3 | 6 - 7 | 817 | 94 | |
GenBank mRNA | AY387697.1 | - | 6 - 7 | 782 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |